ACS, ACS Reagent, Certified ACS
Anhydrous/Dry/Extra Dry
Biochemical
Electronic/Semiconductor
GC, Capillary GC
GC Gradient
GC/Pesticide Residue
Histological
HPLC Gradient
HPLC/UHPLC
Acetonitrile is the simplest organic nitrile. This colorless and polar aprotic solvent is used for organic synthesis and purifications, and to extract fatty acids from oils.
Acetonitrile was first prepared in 1847 by Jean-Baptiste Dumas. In the laboratory, it is commonly used as a medium-polarity solvent. It is miscible with water and some organic solvents, but not with saturated hydrocarbons.
It is popular in cyclic voltammetry, and its ultraviolet transparency, low viscosity, and low chemical reactivity are useful for high-performance liquid chromatography (HPLC). Because it dissolves a range of ionic and nonpolar compounds, acetonitrile serves as a mobile phase solvent in HPLC and LC–MS. It is also the main solvent used to make DNA oligonucleotides from monomers.
Acetonitrile is modestly toxic in small doses. Cases of acetonitrile/cyanide poisoning are rare but can occur by inhalation, ingestion, or skin exposure. Symptoms, which may not appear for several hours after exposure, include respiratory issues, slow pulse rate, nausea, and vomiting. Convulsions and coma can occur and death from respiratory failure may follow.
Synonyms: Methyl cyanide, ethanenitrile (IUPAC name), cyanomethane, ethyl nitrile, methane carbonitrile, MeCN
| Property | Value |
|---|---|
Molecular Weight
|
41.05 g/mol
|
Molar Mass
|
41.053 g/mol
|
Density
|
0.786 g/mL at 25ºC
|
Boiling Point
|
81 to 82ºC
|
Melting Point
|
-48ºC
|
Flash Point
|
5.5ºC
|
Viscosity
|
0.369 cP at 25ºC
|
Dielectric Constant
|
36.64 at 20ºC
|
Dipole Moment
|
36.64 at 20ºC
|
UV Cutoff
|
190 nm
|
Refractive Index
|
1.344 at 20ºC
|
Vapor Pressure
|
72.8 mm Hg (20ºC)
|
Water Solubility
|
24.36 M
|
Heat of Combustion
|
31.03 x 106 J/kg at 25ºC
|
Heat of Vaporization
|
72.7 x 104 J/kg at 80ºC
|
Dissociation Constants
|
pKa = -4.30 (conjugate acid)
|
Surface Tension
|
29.04 dyn/cm at 20ºC
|
Acetonitrile is used as a solvent for the production of pharmaceuticals and photographic film. It is also used to make perfumes, rubber products, pesticides, acrylic nail removers, and batteries.
| Property | Value |
|---|---|
CAS Number
|
75-05-8
|
MDL Number
|
MFCD00001878
|
RIDAD /UN Number
|
UN 1648 3/PG 2
|
Beilstein Registry
|
741857
|
EG/EC Number
|
2008352
|
ACS, ACS Reagent, Certified ACS
Anhydrous/Dry/Extra Dry
Biochemical
Electronic/Semiconductor
GC, Capillary GC
GC Gradient
GC/Pesticide Residue
Histological
HPLC Gradient
HPLC/UHPLC
Laboratory/Technical
LC-MS, Ultra LC-MS
Low Water
Mobile Phase
NF/USP
Purge and Trap
Sequencing
Spectrophotometry
Solvent Blends
Trace Metals