Buy a Mastercycler Nexus Thermal Cycler, Get 5 Cases of PCR Consumables
Purchase an Eppendorf™ Mastercycler™ Nexus Thermal Cycler and get five cases of Eppendorf™ PCR consumables at no extra cost.
Complimentary Products: E0030124260, E0030124286, E0030124812, E0030133374, E951020401
Features and Benefits
The Mastercycler Nexus Thermal Cycler features a universal thermoblock (fits 0.2mL PCR tubes, 96-well PCR plates or 0.5mL PCR tubes); an alternate silver thermoblock allows for faster processing of 0.2mL PCR tubes or 96-well PCR plates. An adjustable-height lid accommodates all PCR consumables.
Eppendorf offers PCR consumables that include 96-well and 384-well PCR plates (for high and medium throughput), divisible plates, PCR tubes and tube strips (for lower throughput), plus sealing options, racks and other accessories.
Qualifying Products
Restrictions
Offer valid until January 31, 2018.
For Healthcare customers, please note that the value of the specific offer(s) the customer may receive from the manufacturer under this program is a discount or other reduction in price to the customer under section 1128B(b)(3)(A) of the Social Security Act [42 U.S.C. 1320a-7b (b) (3)(a)]. Accordingly, the customer shall disclose this and any other discounts or other reduction in price received under this program under any state or federal program which provides cost or charge-based reimbursements to the customer for the products or services purchased under this program. By participating in this promotion, you warrant that you are not a Health Care Practitioner or Health Care Professional as defined in the Code of Massachusetts Regulations, 105 CMR 970, or a healthcare professional as defined under 18.V.S.A. §4631, or a healthcare professional as defined under any similar federal, state or local law regulation.