Buy 1, Get 1 on Broths and Liquid Scintillation Cocktails
2017-nlsu-promo-banner
Purchase one of the MP Biomedicals™ broths or liquid scintillation cocktails listed below and get a second bottle of the same product for no additional cost.
Features and Benefits
MP Biomedicals is well-known for high-quality growth media in various formulations for every molecular biologist. They specialize in formulations for yeast and bacteria and offer more than 1,000 recipes and variations.
Liquid scintillation cocktails from MP Biomedicals include:
  • CytoScint-ES: For one-step filter counting or samples with 0 to 15% water content
  • EcoLume: For samples with moderate to large (40% v/v) water content or high ionic concentrations
  • Ecolite(+): For aqueous sample applications where sample load capacity and enhanced tritium counting efficiency is required
  • Universol-ES: For aqueous samples (0-15%), high ionic concentrations or for controlled chemiluminescence with high tritium efficiency
108-product-image
nlsu-redeem
Download the redemption form (PDF, 36KB)
Qualifying Products
Restrictions
Offer valid until January 31, 2018.
For Healthcare customers, please note that the value of the specific offer(s) the customer may receive from the manufacturer under this program is a discount or other reduction in price to the customer under section 1128B(b)(3)(A) of the Social Security Act [42 U.S.C. 1320a-7b (b) (3)(a)]. Accordingly, the customer shall disclose this and any other discounts or other reduction in price received under this program under any state or federal program which provides cost or charge-based reimbursements to the customer for the products or services purchased under this program. By participating in this promotion, you warrant that you are not a Health Care Practitioner or Health Care Professional as defined in the Code of Massachusetts Regulations, 105 CMR 970, or a healthcare professional as defined under 18.V.S.A. §4631, or a healthcare professional as defined under any similar federal, state or local law regulation.