Buy 1, Get 1 on Applied Biosystems SYBR Green Master Mix
Purchase one Applied Biosystems™ SYBR™ Green Master Mix and get one green master mix at no additional cost.
Features and Benefits
Get everything you need for SYBR Green dye-based PCR amplification and detection in a convenient, single-tube format. Ideal for target identification (screening assays) or when a limited number of assays are needed.
This mix combines SYBR Green I dye, Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial. Premixed components significantly reduce assay setup time. The mix also provides maximum flexibility at a reduced cost because no target-specific Applied Biosystems™ TaqMan™ probes are required.
The SYBR Green I dye chemistry can be used for the following assay types:
- One-step RT-PCR for RNA quantitation
- Two-step RT-PCR for RNA quantitation
- DNA/cDNA quantitation
Qualifying Products
Restrictions
Offer is valid until January 31, 2018. Customers’ eligibility is based on their Fisher Scientific account representative verifying that they are a new lab. Healthcare professionals may not participate in this promotion. By registering, you warrant that you are not prohibited by employment, contract or law from accepting a gift from Fisher Scientific. Offer void where prohibited, licensed or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
For Research Use Only. Not for use in diagnostic procedures.
For Healthcare customers, please note that the value of the specific offer(s) the customer may receive from the manufacturer under this program is a discount or other reduction in price to the customer under section 1128B(b)(3)(A) of the Social Security Act [42 U.S.C. 1320a-7b (b) (3)(a)]. Accordingly, the customer shall disclose this and any other discounts or other reduction in price received under this program under any state or federal program which provides cost or charge-based reimbursements to the customer for the products or services purchased under this program. By participating in this promotion, you warrant that you are not a Health Care Practitioner or Health Care Professional as defined in the Code of Massachusetts Regulations, 105 CMR 970, or a healthcare professional as defined under 18.V.S.A. §4631, or a healthcare professional as defined under any similar federal, state or local law regulation.