Buy 2, Get 1 on TaqMan Fast Advance Master Mix
2017-nlsu-promo-banner
Purchase two Applied Biosystems™ TaqMan™ Fast Advance Master Mix or Universal Master Mix II without UNG products and get one master mix product at no additional cost.
Features and Benefits
TaqMan Fast Advanced Master Mix has been designed to match or exceed the performance of standard master mixes, delivering shorter run times (<40 minutes) with results equal to or better than what is achieved today. The mix contains Applied Biosystems™ AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs with dUTP, Applied Biosystems™ ROX™ dye (passive reference), and optimized buffer components. 
  • Supplied as 2X concentrate
  • Optimized for multiplexing
  • Engineered for enhanced benchtop stability
  • Reduced run times on fast and standard instrumentation
TaqMan Universal Master Mix II without UNG is a convenient mix of components needed to perform real-time PCR. Use it to amplify complementary DNA (cDNA) and other DNA targets for a variety of applications, including quantitation and genotyping. The mix contains Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure), dNTPs with dUTP, Applied Biosystems™ ROX™ Passive Reference, and optimized buffer components.
  • Supplied as 2X concentrate
  • Optimized for use with primers and TaqMan probes
  • Guaranteed minimum shelf life: 60 days
111-product-image1
nlsu-redeem
Download the redemption form (PDF, 36KB)
Qualifying Products
Restrictions
Offer is valid until January 31, 2018. Customers’ eligibility is based on their Fisher Scientific account representative verifying that they are a new lab. Healthcare professionals may not participate in this promotion. By registering, you warrant that you are not prohibited by employment, contract or law from accepting a gift from Fisher Scientific. Offer void where prohibited, licensed or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
For Research Use Only. Not for use in diagnostic procedures.
For Healthcare customers, please note that the value of the specific offer(s) the customer may receive from the manufacturer under this program is a discount or other reduction in price to the customer under section 1128B(b)(3)(A) of the Social Security Act [42 U.S.C. 1320a-7b (b) (3)(a)]. Accordingly, the customer shall disclose this and any other discounts or other reduction in price received under this program under any state or federal program which provides cost or charge-based reimbursements to the customer for the products or services purchased under this program. By participating in this promotion, you warrant that you are not a Health Care Practitioner or Health Care Professional as defined in the Code of Massachusetts Regulations, 105 CMR 970, or a healthcare professional as defined under 18.V.S.A. §4631, or a healthcare professional as defined under any similar federal, state or local law regulation.