Get a Saddle Stool with Biological Safety Cabinet Purchase
When you buy a qualifying Thermo Scientific™ 1300 Series Class II, Type A2 Biological Safety Cabinet (BSC) Package, you’ll get a Thermo Scientific™ Ergolign™ Saddle Stool at no additional cost. Once you’ve made your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Thermo Scientific BSCs are designed to protect researchers from contaminated materials and samples. Equipped with Thermo Scientific™ SmartFlow™ technology, they feature dual-DC motors that automatically balance the cabinet inflow and downflow velocities in real time, even as the filters load. For added confidence, Digital Airflow Verification (DAVe) technology signals if conditions exceed predetermined limits.
| Catalog Number | Description | ||
|---|---|---|---|
Thermo Scientific Ergolign Saddle Stool, 23 in. | Learn More |