Get a Disposal Box with Fisherbrand Tubes Purchase
When you buy three cases of any Fisherbrand™ Round Bottom Disposable Borosilicate Glass Tubes with Plain End, you’ll get a pack of Fisherbrand glass disposal boxes at no additional cost.
Features and Benefits
Fisherbrand Disposable Borosilicate Glass Tubes with Plain End are made to reduce pH changes and contaminants potentially leached from soda lime glass. Use these tubes for tissue culture and clinical chemistry applications. Keep glass separate from other waste with Fisherbrand glass disposal boxes. They’re manufactured from sturdy corrugated cardboard and come with polyethylene liner bags.
Catalog Number | Description | ||
---|---|---|---|
Fisherbrand Glass-Disposal Boxes | Learn More |