Assay Details
Gene Family ID: | MIPF0000196, mir-335 |
Mature miRNA Sequence: | UCAAGAGCAAUAACGAAAAAUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-335-5p | MIMAT0000765 | |
| Mouse | mmu-miR-335-5p | MIMAT0000766 | |
| Rat | rno-miR-335 | MIMAT0000575 | |
| Bovine | bta-miR-335 | MIMAT0009291 | |
| Dog | cfa-miR-335 | MIMAT0006624 | |
| Eptesicus fuscus | efu-miR-335 | MIMAT0035059 | |
| View More Rows | |||
| Horse | eca-miR-335 | MIMAT0012942 | |
| Pan troglodytes | ptr-miR-335 | MIMAT0008104 | |
| Pongo pygmaeus | ppy-miR-335 | MIMAT0015837 | |
| Rhesus monkey | mml-miR-335-5p | MIMAT0006274 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Eptesicus fuscus
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-335 (v17)
Mouse: mmu-miR-335 (v9.2)
Macaca mulatta: mml-miR-335 (v19)
Mouse: mmu-miR-335 (v9.2)
Macaca mulatta: mml-miR-335 (v19)

Genomic Map