Assay Details
Gene Family ID: | MIPF0000196, mir-335 |
Mature miRNA Sequence: | UCAAGAGCAAUAACGAAAAAUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-335-5p | MIMAT0000765 |
| Mouse | mmu-miR-335-5p | MIMAT0000766 |
| Rat | rno-miR-335 | MIMAT0000575 |
| Bovine | bta-miR-335 | MIMAT0009291 |
| Dog | cfa-miR-335 | MIMAT0006624 |
| Eptesicus fuscus | efu-miR-335 | MIMAT0035059 |
| View More Rows | ||
| Horse | eca-miR-335 | MIMAT0012942 |
| Pan troglodytes | ptr-miR-335 | MIMAT0008104 |
| Pongo pygmaeus | ppy-miR-335 | MIMAT0015837 |
| Rhesus monkey | mml-miR-335-5p | MIMAT0006274 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Eptesicus fuscus
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-335 (v17)
Mouse: mmu-miR-335 (v9.2)
Macaca mulatta: mml-miR-335 (v19)
Mouse: mmu-miR-335 (v9.2)
Macaca mulatta: mml-miR-335 (v19)

Genomic Map