Assay Details
| Gene Family ID: | MIPF0000196, mir-335 | 
| Mature miRNA Sequence: | UCAAGAGCAAUAACGAAAAAUGU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-335-5p | MIMAT0000765 | 
| Mouse | mmu-miR-335-5p | MIMAT0000766 | 
| Rat | rno-miR-335 | MIMAT0000575 | 
| Bovine | bta-miR-335 | MIMAT0009291 | 
| Dog | cfa-miR-335 | MIMAT0006624 | 
| Eptesicus fuscus | efu-miR-335 | MIMAT0035059 | 
| View More Rows | ||
| Horse | eca-miR-335 | MIMAT0012942 | 
| Pan troglodytes | ptr-miR-335 | MIMAT0008104 | 
| Pongo pygmaeus | ppy-miR-335 | MIMAT0015837 | 
| Rhesus monkey | mml-miR-335-5p | MIMAT0006274 | 
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Eptesicus fuscus
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
 Back To Top
 Back To TopMore Information
Related Products
miRBase Alias:
						
							Human: hsa-miR-335 (v17)
Mouse: mmu-miR-335 (v9.2)
Macaca mulatta: mml-miR-335 (v19)
						
					
				Mouse: mmu-miR-335 (v9.2)
Macaca mulatta: mml-miR-335 (v19)
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :