Assay Details
Gene Family ID: | MIPF0000029, mir-133 |
Mature miRNA Sequence: | UUGGUCCCCUUCAACCAGCUA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Brugia malayi | bma-miR-133 | MIMAT0014117 |
Chicken | gga-miR-133b | MIMAT0001138 |
Ciona savignyi | csa-miR-133 | MIMAT0006128 |
Xenopus laevis | xla-miR-133b | MIMAT0011137 |
Xenopus tropicalis | xtr-miR-133b | MIMAT0003597 |
Stem-loop Details
Brugia malayi
Chicken
Ciona savignyi
Xenopus laevis
Xenopus tropicalis

More Information
Related Products
miRBase Alias:
Human: hsa-miR-133b (v9.2)
Mouse: mmu-miR-133b (v9.2)
Rat: rno-miR-133b (v9.2)
Mouse: mmu-miR-133b (v9.2)
Rat: rno-miR-133b (v9.2)
