Assay Details
Gene Family ID: | MIPF0000029, mir-133 |
Mature miRNA Sequence: | UUGGUCCCCUUCAACCAGCUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Brugia malayi | bma-miR-133 | MIMAT0014117 |
| Chicken | gga-miR-133b | MIMAT0001138 |
| Ciona savignyi | csa-miR-133 | MIMAT0006128 |
| Xenopus laevis | xla-miR-133b | MIMAT0011137 |
| Xenopus tropicalis | xtr-miR-133b | MIMAT0003597 |
Stem-loop Details
Brugia malayi
Chicken
Ciona savignyi
Xenopus laevis
Xenopus tropicalis

Genomic Map