Assay Details
Gene Family ID: | MIPF0000189, mir-345 |
Mature miRNA Sequence: | UGCUGACCCCUAGUCCAGUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-345-5p | MIMAT0000594 |
Stem-loop Details
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-345 (v9.2)
Rat: rno-miR-345 (v9.2)
Rat: rno-miR-345 (v9.2)

Genomic Map