Assay Details
Amplicon Length: | 65 |
Chromosome Location: | Chr. 6 - 132748818 - 132748818 [+] on 132748818 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-345-5p | MIMAT0000594 |
| Rat | rno-miR-345-3p | MIMAT0004655 |
Stem-loop Details
Rat

Genomic Map