Assay Details
Mature miRNA Sequence: | GGUUCGUACGUACACUGUUCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR416 | MIMAT0001324 |
Stem-loop Details
Arabidopsis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ath-miR416 |
|---|---|
| miRBase Accession Number: | MI0001427 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGUUCGUACGUACACUGUUCA |
| Species: | Arabidopsis |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | GGUUCGUACGUACACUGUUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR416 | MIMAT0001324 |