Assay Details
| Mature miRNA Sequence: | GGUUCGUACGUACACUGUUCA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Arabidopsis | ath-miR416 | MIMAT0001324 | 
Stem-loop Details
Arabidopsis
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ath-miR416 | 
|---|---|
| miRBase Accession Number: | MI0001427 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | GGUUCGUACGUACACUGUUCA | 
| Species: | Arabidopsis | 
| Product Type: | mirVana® miRNA mimic | 
| Mature miRNA Sequence: | GGUUCGUACGUACACUGUUCA | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Arabidopsis | ath-miR416 | MIMAT0001324 | 
 Back To Top
 Back To Top