Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCUAGAGGGAAGCGCUUUCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-518e-5p | MIMAT0005450 | |
| Human | hsa-miR-519a-5p | MIMAT0005452 | |
| Human | hsa-miR-519b-5p | MIMAT0005454 | |
| Human | hsa-miR-519c-5p | MIMAT0002831 | |
| Human | hsa-miR-522-5p | MIMAT0005451 | |
| Human | hsa-miR-523-5p | MIMAT0005449 | |
| View More Rows | |||
| Gorilla gorilla | ggo-miR-518e-5p | MIMAT0036427 | |
| Gorilla gorilla | ggo-miR-519a-5p | MIMAT0036439 | |
| Gorilla gorilla | ggo-miR-519c-5p | MIMAT0036410 | |
| Gorilla gorilla | ggo-miR-519c | MIMAT0036435 | |
| Gorilla gorilla | ggo-miR-522-5p | MIMAT0036437 | |
| Gorilla gorilla | ggo-miR-523-5p | MIMAT0036414 | |
| Pan troglodytes | ptr-miR-519f-5p | MIMAT0036456 | |
| Rhesus monkey | mml-miR-519a | MIMAT0036445 | |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-518e* (v17)
Human: hsa-miR-522* (v17)
Human: hsa-miR-523* (v17)
Human: hsa-miR-519a* (v17)
Pongo pygmaeus: ppy-miR-519c-5p (v21)
Pongo pygmaeus: ppy-miR-519b-5p (v21)
Human: hsa-miR-522* (v17)
Human: hsa-miR-523* (v17)
Human: hsa-miR-519a* (v17)
Pongo pygmaeus: ppy-miR-519c-5p (v21)
Pongo pygmaeus: ppy-miR-519b-5p (v21)

Genomic Map