Assay Details
Amplicon Length: | 135 |
Chromosome Location: | Chr. 19 - 53695468 - 53695468 [-] on 53695468 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519b-5p | MIMAT0005454 |
| Human | hsa-miR-519b-3p | MIMAT0002837 |
Stem-loop Details
Human

Genomic Map