Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCAUCCUUUUAGAGGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519b-3p | MIMAT0002837 |
| Gorilla gorilla | ggo-miR-519c-3p | MIMAT0036411 |
| Pan troglodytes | ptr-miR-519b | MIMAT0008193 |
| Pongo pygmaeus | ppy-miR-519b | MIMAT0015959 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
More Information
Related Products
miRBase Alias:
Pongo pygmaeus: ppy-miR-519b-3p (v21)
Pongo pygmaeus: ppy-miR-519a (v21)
Pongo pygmaeus: ppy-miR-519a (v21)

Genomic Map