Assay Details
Gene Family ID: | MIPF0000212, mir-540 |
Mature miRNA Sequence: | AGGUCAGAGGUCGAUCCUGGGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-540-3p | MIMAT0003174 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-540 |
|---|---|
| miRBase Accession Number: | MI0003524 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUCAGAGGUCGAUCCUGGGC |
| Species: | Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000212, mir-540 |
Mature miRNA Sequence: | AGGUCAGAGGUCGAUCCUGGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-540-3p | MIMAT0003174 |