Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 6 - 133706457 - 133706457 [+] on 133706457 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-540-5p | MIMAT0017211 |
| Rat | rno-miR-540-3p | MIMAT0003174 |
Stem-loop Details
Rat

Genomic Map