Assay Details
Mature miRNA Sequence: | UACUGGCCUGUUAAGUCCCAAGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Tribolium castaneum | tca-miR-193-3p | MIMAT0018897 |
Stem-loop Details
Tribolium castaneum

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | tca-miR-193-3p |
---|---|
miRBase Accession Number: | MI0016396 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UACUGGCCUGUUAAGUCCCAAGU |
Species: | Tribolium castaneum |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UACUGGCCUGUUAAGUCCCAAGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Tribolium castaneum | tca-miR-193-3p | MIMAT0018897 |