Assay Details
Mature miRNA Sequence: | UACUGGCCUGUUAAGUCCCAAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tribolium castaneum | tca-miR-193-3p | MIMAT0018897 |
Stem-loop Details
Tribolium castaneum
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | tca-miR-193-3p |
|---|---|
| miRBase Accession Number: | MI0016396 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUGGCCUGUUAAGUCCCAAGU |
| Species: | Tribolium castaneum |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UACUGGCCUGUUAAGUCCCAAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tribolium castaneum | tca-miR-193-3p | MIMAT0018897 |