Assay Details
Gene Family ID: | MIPF0000725, mir-1287 |
Mature miRNA Sequence: | CUCUAGCCACAGAUGCAGUGAU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1287-3p | MIMAT0026738 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-1287-3p |
---|---|
miRBase Accession Number: | MI0006349 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CUCUAGCCACAGAUGCAGUGAU |
Species: | Human |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000725, mir-1287 |
Mature miRNA Sequence: | CUCUAGCCACAGAUGCAGUGAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1287-3p | MIMAT0026738 |