Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 10 - 98395518 - 98395518 [-] on 98395518 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1287-5p | MIMAT0005878 |
Human | hsa-miR-1287-3p | MIMAT0026738 |
Stem-loop Details
Human
