Assay Details
Gene Family ID: | MIPF0000437, mir-504 |
Mature miRNA Sequence: | GGGAGUGCAGGGCAGGGUUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-504-3p | MIMAT0026612 |
| Pteropus alecto | pal-miR-504-3p | MIMAT0040012 |
Stem-loop Details
Human
Pteropus alecto

Genomic Map