Assay Details
Amplicon Length: | 105 |
Chromosome Location: | Chr. X - 138668201 - 138668201 [+] on 138668201 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-504-5p | MIMAT0002875 |
| Human | hsa-miR-504-3p | MIMAT0026612 |
Stem-loop Details
Human

Genomic Map