Assay Details
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | CGGGGCCGUAGCACUGUCUGAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-128-1-5p | MIMAT0026477 | |
| Cricetulus griseus | cgr-miR-128-5p | MIMAT0023749 | |
| Dasypus novemcinctus | dno-miR-128-5p | MIMAT0047621 | |
| Goat | chi-miR-128-5p | MIMAT0035933 | |
| Guinea pig | cpo-miR-128-5p | MIMAT0046974 | |
| Monodelphis domestica | mdo-miR-128b-5p | MIMAT0028608 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-128-5p | MIMAT0039793 | |
| Rabbit | ocu-miR-128a-5p | MIMAT0048210 | |
| Rhesus monkey | mml-miR-128a-5p | MIMAT0026565 | |
Stem-loop Details
Human
Cricetulus griseus
Dasypus novemcinctus
Goat
Guinea pig
Monodelphis domestica
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map