Assay Details
| Gene Family ID: | MIPF0000048, mir-128 | 
| Mature miRNA Sequence: | CGGGGCCGUAGCACUGUCUGAGA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-128-1-5p | MIMAT0026477 | 
| Cricetulus griseus | cgr-miR-128-5p | MIMAT0023749 | 
| Dasypus novemcinctus | dno-miR-128-5p | MIMAT0047621 | 
| Goat | chi-miR-128-5p | MIMAT0035933 | 
| Guinea pig | cpo-miR-128-5p | MIMAT0046974 | 
| Monodelphis domestica | mdo-miR-128b-5p | MIMAT0028608 | 
| View More Rows | ||
| Pteropus alecto | pal-miR-128-5p | MIMAT0039793 | 
| Rabbit | ocu-miR-128a-5p | MIMAT0048210 | 
| Rhesus monkey | mml-miR-128a-5p | MIMAT0026565 | 
Stem-loop Details
Human
Cricetulus griseus
Dasypus novemcinctus
Goat
Guinea pig
Monodelphis domestica
Pteropus alecto
Rabbit
Rhesus monkey

 Genomic Map
Genomic Map