Assay Details
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | GGGGGCCGAUACACUGUACGAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-128-2-5p | MIMAT0031095 |
| Dasypus novemcinctus | dno-miR-128-2-5p | MIMAT0047623 |
| Guinea pig | cpo-miR-128-2-5p | MIMAT0046976 |
| Rabbit | ocu-miR-128b-5p | MIMAT0048212 |
| Rhesus monkey | mml-miR-128b-5p | MIMAT0026814 |
Stem-loop Details
Human
Dasypus novemcinctus
Guinea pig
Rabbit
Rhesus monkey

Genomic Map