Assay Details
| Gene Family ID: | MIPF0000048, mir-128 | 
| Mature miRNA Sequence: | GGGGGCCGAUACACUGUACGAGA | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-128-2-5p | MIMAT0031095 | 
| Dasypus novemcinctus | dno-miR-128-2-5p | MIMAT0047623 | 
| Guinea pig | cpo-miR-128-2-5p | MIMAT0046976 | 
| Rabbit | ocu-miR-128b-5p | MIMAT0048212 | 
| Rhesus monkey | mml-miR-128b-5p | MIMAT0026814 | 
Stem-loop Details
Human
Dasypus novemcinctus
Guinea pig
Rabbit
Rhesus monkey
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :