Assay Details
Gene Family ID: | MIPF0001796, mir-2070 |
Mature miRNA Sequence: | AUGUGCAUAAGCUGUGGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2070-3p | MIMAT0031637 |
Stem-loop Details
Branchiostoma belcheri

Genomic Map