Assay Details
Gene Family ID: | MIPF0001796, mir-2070 |
Mature miRNA Sequence: | AUGUGCAUAAGCUGUGGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2070-3p | MIMAT0031637 |
Stem-loop Details
Branchiostoma belcheri
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bbe-miR-2070-3p |
|---|---|
| miRBase Accession Number: | MI0026215 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUGUGCAUAAGCUGUGGGA |
| Species: | Branchiostoma belcheri |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001796, mir-2070 |
Mature miRNA Sequence: | AUGUGCAUAAGCUGUGGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2070-3p | MIMAT0031637 |