Assay Details
Gene Family ID: | MIPF0001349, mir-3544 |
Mature miRNA Sequence: | AGAAAGGCAUCAUAUAGGAGCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3544-5p | MIMAT0022353 |
| Callithrix jacchus | cja-miR-3544 | MIMAT0039692 |
Stem-loop Details
Mouse
Callithrix jacchus

Genomic Map