Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 12 - 109585631 - 109585631 [-] on 109585631 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-337-3p | MIMAT0000578 |
| Mouse | mmu-miR-337-5p | MIMAT0004644 |
| Mouse | mmu-miR-3544-5p | MIMAT0022353 |
| Mouse | mmu-miR-3544-3p | MIMAT0022354 |
| Mouse | mmu-miR-540-5p | MIMAT0004786 |
| Mouse | mmu-miR-540-3p | MIMAT0003167 |
Stem-loop Details
Mouse

Genomic Map