Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UAGGUAGUUUCCUGUUGUUGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-196b-5p | MIMAT0001080 |
Mouse | mmu-miR-196b-5p | MIMAT0001081 |
Rat | rno-miR-196b-5p | MIMAT0001082 |
Callithrix jacchus | cja-miR-196b | MIMAT0039444 |
Cricetulus griseus | cgr-miR-196b | MIMAT0023833 |
Gorilla gorilla | ggo-miR-196b | MIMAT0049110 |
View More Rows | ||
Horse | eca-miR-196b | MIMAT0012939 |
Microcebus murinus | mmr-miR-196b | MIMAT0049222 |
Monodelphis domestica | mdo-miR-196a-5p | MIMAT0028494 |
Nomascus leucogenys | nle-miR-196b | MIMAT0049406 |
Otolemur garnettii | oga-miR-196b | MIMAT0049655 |
Pan troglodytes | ptr-miR-196b | MIMAT0008061 |
Pig | ssc-miR-196b-5p | MIMAT0013923 |
Pongo pygmaeus | ppy-miR-196b | MIMAT0015791 |
Pteropus alecto | pal-miR-196b-5p | MIMAT0040009 |
Rhesus monkey | mml-miR-196b-5p | MIMAT0006229 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Gorilla gorilla
Horse
Microcebus murinus
Monodelphis domestica
Nomascus leucogenys
Otolemur garnettii
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
