Assay Details
Mature miRNA Sequence: | CCAGGGAGGCUGGUUUGGAGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6754-5p | MIMAT0027408 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6754-5p |
---|---|
Stem-loop Accession Number: | MI0022599 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CCAGGGAGGCUGGUUUGGAGGA |
Species: | Human |
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CCAGGGAGGCUGGUUUGGAGGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6754-5p | MIMAT0027408 |