Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 11 - 71473725 - 71473725 [+] on 71473725 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6754-3p | MIMAT0027409 |
Human | hsa-miR-6754-5p | MIMAT0027408 |
Stem-loop Details
Human
