Assay Details
Gene Family ID: | MIPF0000838, mir-2240 |
Mature miRNA Sequence: | UCACCGGGAUUCUCAGUGGAUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2240a | MIMAT0011690 |
Stem-loop Details
Pristionchus pacificus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppc-miR-2240a |
|---|---|
| miRBase Accession Number: | MI0011190 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACCGGGAUUCUCAGUGGAUU |
| Species: | Pristionchus pacificus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000838, mir-2240 |
Mature miRNA Sequence: | UCACCGGGAUUCUCAGUGGAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2240a | MIMAT0011690 |