Assay Details
Gene Family ID: | MIPF0000838, mir-2240 |
Mature miRNA Sequence: | UCACCGGGAUUCUCAGUGGAUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pristionchus pacificus | ppc-miR-2240a | MIMAT0011690 |
Stem-loop Details
Pristionchus pacificus
