Assay Details
Gene Family ID: | MIPF0002097, mir-3543 |
Mature miRNA Sequence: | CAGGAGUCGAGUGAUGGUUCAAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3543 | MIMAT0017797 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-3543 |
|---|---|
| miRBase Accession Number: | MI0015397 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGGAGUCGAGUGAUGGUUCAAA |
| Species: | Rat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0002097, mir-3543 |
Mature miRNA Sequence: | CAGGAGUCGAGUGAUGGUUCAAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3543 | MIMAT0017797 |