Assay Details
Amplicon Length: | 52 |
Chromosome Location: | Chr. 6 - 133716464 - 133716464 [-] on 133716464 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3543 | MIMAT0017797 |
| Rat | rno-miR-434-3p | MIMAT0005315 |
| Rat | rno-miR-434-5p | MIMAT0017307 |
Stem-loop Details
Rat

Genomic Map