Assay Details
Gene Family ID: | MIPF0000195, mir-337 |
Mature miRNA Sequence: | UCAGCUCCUAUAUGAUGCCUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-337-3p | MIMAT0000578 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-337-3p |
|---|---|
| miRBase Accession Number: | MI0000615 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAGCUCCUAUAUGAUGCCUUU |
| Species: | Mouse |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000195, mir-337 |
Mature miRNA Sequence: | UCAGCUCCUAUAUGAUGCCUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-337-3p | MIMAT0000578 |