Assay Details
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UGGUAAACUUUGUUCAGUUUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila virilis | dvi-miR-309-5p | MIMAT0032081 |
Stem-loop Details
Drosophila virilis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dvi-miR-309-5p |
|---|---|
| miRBase Accession Number: | MI0009526 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUAAACUUUGUUCAGUUUUG |
| Species: | Drosophila virilis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UGGUAAACUUUGUUCAGUUUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila virilis | dvi-miR-309-5p | MIMAT0032081 |