Assay Details
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UGGUAAACUUUGUUCAGUUUUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila virilis | dvi-miR-309-5p | MIMAT0032081 |
Stem-loop Details
Drosophila virilis

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | dvi-miR-309-5p |
---|---|
miRBase Accession Number: | MI0009526 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGGUAAACUUUGUUCAGUUUUG |
Species: | Drosophila virilis |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UGGUAAACUUUGUUCAGUUUUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila virilis | dvi-miR-309-5p | MIMAT0032081 |