Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | GCGUCGAGCCCCACGCACAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ophiophagus hannah | oha-miR-375-5p | MIMAT0036917 |
Stem-loop Details
Ophiophagus hannah

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | oha-miR-375-5p |
---|---|
miRBase Accession Number: | MI0031466 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GCGUCGAGCCCCACGCACAA |
Species: | Ophiophagus hannah |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | GCGUCGAGCCCCACGCACAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ophiophagus hannah | oha-miR-375-5p | MIMAT0036917 |