Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | GCGUCGAGCCCCACGCACAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-375-5p | MIMAT0036917 |
Stem-loop Details
Ophiophagus hannah
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | oha-miR-375-5p |
|---|---|
| miRBase Accession Number: | MI0031466 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCGUCGAGCCCCACGCACAA |
| Species: | Ophiophagus hannah |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | GCGUCGAGCCCCACGCACAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-375-5p | MIMAT0036917 |