Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | CUAUGUAACAUGGUCCACUAAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-379-3p | MIMAT0004791 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-379-3p |
|---|---|
| miRBase Accession Number: | MI0003541 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUGUAACAUGGUCCACUAAC |
| Species: | Rat |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | CUAUGUAACAUGGUCCACUAAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-379-3p | MIMAT0004791 |