Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 6 - 133858143 - 133858143 [-] on 133858143 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-379-5p | MIMAT0003192 |
| Rat | rno-miR-379-3p | MIMAT0004791 |
Stem-loop Details
Rat

Genomic Map