Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. X - 7242382 - 7242382 [-] on 7242382 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-362-3p | MIMAT0004684 |
| Mouse | mmu-miR-362-5p | MIMAT0000706 |
Stem-loop Details
Mouse

Genomic Map